N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 95%

Supplier: Thermo Fisher Scientific

H52181.MD H52181.03 H52181.06
H52181.MDEA 38.8 EUR
H52181.MD H52181.03 H52181.06
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 95%
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine

Purity: 95% CAS No.: 57292-44-1 Molecular Formula: C14H18ClNO4 Formula Weight: 299.75 Melting Point: 110° Beilstein Reference: 5381988 Optical Rotation: -26° (c=1 in ethyl acetate) MDL No.: MFCD00076978

Formula: C₁₄H₁₈ClNO₄
Melting Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00076978
CAS Number: 57292-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR