2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

H52312.MD H52312.03
H52312.MDEA 97.8 EUR
H52312.MD H52312.03
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester 96%
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1073338-92-7 Molecular Formula: C13H19BO5 Formula Weight: 266.10

Formula: C₁₃H₁₉BO₅
Storage Temperature: Ambient
MDL Number: MFCD11855980
CAS Number: 1073338-92-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR