Ethyl-4-(1-piperazinyl)benzoate ≥97%

Supplier: Thermo Fisher Scientific
Warning

H52400.MD H52400.03
H52400.MDEA 0 EUR
H52400.MD H52400.03
Ethyl-4-(1-piperazinyl)benzoate ≥97%
Ethyl-4-(1-piperazinyl)benzoate

Purity: 97+% CAS No.: 80518-57-6 Molecular Formula: C13H18N2O2 Formula Weight: 234.30 MDL No.: MFCD04973340

Formula: C₁₃H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04973340
CAS Number: 80518-57-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR