tert-Butyl 2-phenylpiperazine-1-carboxylate 97%

Supplier: Thermo Fisher Scientific
Warning

H52797.MD H52797.03
H52797.MDEA 127 EUR
H52797.MD H52797.03
tert-Butyl 2-phenylpiperazine-1-carboxylate 97%
tert-Butyl 2-phenylpiperazine-1-carboxylate

Purity: 97% CAS No.: 886766-60-5 Molecular Formula: C15H22N2O2 Formula Weight: 262.35 MDL No.: MFCD07367734

Formula: C₁₅H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07367734
CAS Number: 886766-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR