(S)-3-Hydroxypyrrolidine hydrochloride 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3S)-pyrrolidin-3-ol hydrochloride

H52815.03 H52815.06
H52815.03EA 37.8 EUR
H52815.03 H52815.06
(S)-3-Hydroxypyrrolidine hydrochloride 97%
(S)-3-Hydroxypyrrolidine hydrochloride

Purity: 97% CAS No.: 122536-94-1 Molecular Formula: C4H9NO.HCl Formula Weight: 123.58 Melting Point: 106-107° Boiling Point: 108-110°/8mm MDL No.: MFCD00272298

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
Boiling Pt: 108…110 °C (8 mmHg)
Melting Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00272298
CAS Number: 122536-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR