3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%

Supplier: Thermo Fisher Scientific
Warning

H53484.03 H53484.06 H53484.14
H53484.03EA 60.3 EUR
H53484.03 H53484.06 H53484.14
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine

Purity: 96% CAS No.: 1671-87-0 Molecular Formula: C12H8N6 Formula Weight: 236.24 Melting Point: 225° dec. MDL No.: MFCD00121717

Formula: C₁₂H₈N₆
Melting Pt: 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00121717
CAS Number: 1671-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR