Pentafluorophenyl trifluoroacetate ≥98%

Supplier: Thermo Fisher Scientific
Warning

H54529.14 H54529.06
H54529.14EA 340 EUR
H54529.14 H54529.06
Pentafluorophenyl trifluoroacetate ≥98%
Pentafluorophenyl trifluoroacetate

Purity: 98+% CAS No.: 14533-84-7 Molecular Formula: C8F8O2 Formula Weight: 280.07 Flash Point: 52°(126°F) Refractive Index: 1.368 Boiling Point: 122-123° Beilstein Reference: 2003848 UN No.: UN3272 MDL No.: MFCD00134438

Formula: C₈F₈O₂
Boiling Pt: 122…123 °C
Density: 1,63
Flash Pt: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134438
CAS Number: 14533-84-7
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR