1H-Indole-3-carbaldehyde oxime 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1H-Indole-3-carbaldehyde Oxime, 1H-Indole-3-carboxaldehyde Oxime

H54754.03
H54754.03EA 0 EUR
H54754.03
1H-Indole-3-carbaldehyde oxime 97%
1H-Indole-3-carbaldehyde oxime

Purity: 97% CAS No.: 2592-05-4 Molecular Formula: C9H8N2O Formula Weight: 160.17 MDL No.: MFCD00022737

Formula: C₉H₈N₂O
MW: 160.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022737
CAS Number: 2592-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR