4-Fluoro-3-methoxyphenylboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

H54778.MD H54778.06
H54778.MDEA 0 EUR
H54778.MD H54778.06
4-Fluoro-3-methoxyphenylboronic acid pinacol ester 96%
4-Fluoro-3-methoxyphenylboronic acid pinacol ester

Purity: 96% CAS No.: 425378-85-4 Molecular Formula: C13H18BFO3 Formula Weight: 252.09 Melting Point: 69-71° MDL No.: MFCD11227055

Formula: C₁₃H₁₈BFO₃
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD11227055
CAS Number: 425378-85-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR