2-Methyl-7-quinolineboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific
Warning

H54984.MDEA 0 EUR
H54984.MD
2-Methyl-7-quinolineboronic acid pinacol ester 96%
2-Methyl-7-quinolineboronic acid pinacol ester

Purity: 96% Molecular Formula: C16H20BNO2 Formula Weight: 269.15 Storage Temperature: 2°C to 8°C Melting Point: 100-102° MDL No.: MFCD11846348

Formula: C₁₆H₂₀BNO₂
Melting Pt: 100…102 °C
MDL Number: MFCD11846348
CAS Number: 939052-79-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR