4-Bromo-N-methylaniline ≥97%

Supplier: Thermo Fisher Scientific
Warning

H55273.03 H55273.06
H55273.03EA 63.5 EUR
H55273.03 H55273.06
4-Bromo-N-methylaniline ≥97%
4-Bromo-N-methylaniline
Formula: C₇H₈BrN
Boiling Pt: 259…260 °C
Density: 1,482
Storage Temperature: Ambient
MDL Number: MFCD03030391
CAS Number: 6911-87-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR