Triphenylantimony dichloride ≥99%

Supplier: Thermo Fisher Scientific
Danger

H55719.06 H55719.14
H55719.06EA 67.9 EUR
H55719.06 H55719.14
Triphenylantimony dichloride ≥99%
Triphenylantimony dichloride
Formula: C₁₈H₁₅Cl₂Sb
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00013598
CAS Number: 594-31-0
EINECS: 209-834-1
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR