Pentacene 97

Supplier: Thermo Fisher Scientific

Synonyms: 2,3,6,7-Dibenzanthracene

H56310.MD H56310.03
H56310.MDEA 157 EUR
H56310.MD H56310.03
Pentacene 97
Pentacene
Formula: C₂₂H₁₄
MW: 278.35 g/mol
Melting Pt: 372…374 °C (subl.)
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003710
CAS Number: 135-48-8
EINECS: 205-193-7
Merck Index: 14,7107

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR