(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥97%

Supplier: Thermo Fisher Scientific
Warning

H57217.03
H57217.03EA 0 EUR
H57217.03
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥97%
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate

Purity: 97+% CAS No.: 138108-72-2 Molecular Formula: C10H19NO3 Formula Weight: 201.27 MDL No.: MFCD03094725

Formula: C₁₀H₁₉NO₃
Density: 1,089
Storage Temperature: Ambient
MDL Number: MFCD03094725
CAS Number: 138108-72-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR