tert-Butyl methyl malonate 95%

Supplier: Thermo Fisher Scientific
Warning

H59540.AB
H59540.ABEA 112 EUR
H59540.AB
tert-Butyl methyl malonate 95%
tert-Butyl methyl malonate
Formula: C₈H₁₄O₄
Boiling Pt: 80 °C (11 mmHg)
Density: 1,030
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042856
CAS Number: 42726-73-8
EINECS: 255-919-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR