2-Quinoxalinecarboxylic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

H59780.MEEA 145 EUR
H59780.ME
2-Quinoxalinecarboxylic acid ≥97%
2-Quinoxalinecarboxylic acid
Formula: C₉H₆N₂O₂
Melting Pt: 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012334
CAS Number: 879-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR