Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) ≥98%

Supplier: Thermo Fisher Scientific
Warning

H60442.MC H60442.36 H60442.22
H60442.MCEA 0 EUR
H60442.MC H60442.36 H60442.22
Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) ≥98%
Bis(α,α-dimethylbenzyl) peroxide (Dicumyl peroxide)
Formula: C₁₈H₂₂O₂
Melting Pt: 39…41 °C
Density: 1,56
Storage Temperature: Refrigerator
MDL Number: MFCD00036227
CAS Number: 80-43-3
EINECS: 201-279-3
UN: 3110
ADR: 5.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR