4-Bromo-2,6-difluorobenzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Bromo-2,6-difluorophenyl)methanol

H61623.03 H61623.06
H61623.03EA 44.7 EUR
H61623.03 H61623.06
4-Bromo-2,6-difluorobenzyl alcohol ≥97%
4-Bromo-2,6-difluorobenzyl alcohol
Formula: C₇H₅BrF₂O
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03094461
CAS Number: 162744-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR