(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%

Supplier: Thermo Fisher Scientific

H61638.03 H61638.06
H61638.03EA 83.3 EUR
H61638.03 H61638.06
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate

Purity: 98% CAS No.: 93267-04-0 Molecular Formula: C9H16INO4 Formula Weight: 329.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White Melting Point: 50-52° Beilstein Reference: 4422011 MDL No.: MFCD00216579

Formula: C₉H₁₆INO₄
Melting Pt: 50…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216579
CAS Number: 93267-04-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR