3-Cyano-2-fluorophenylboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

H62113.03
H62113.03EA 0 EUR
H62113.03
3-Cyano-2-fluorophenylboronic acid pinacol ester 96%
3-Cyano-2-fluorophenylboronic acid pinacol ester

Purity: 96% CAS No.: 1218791-15-1 Molecular Formula: C13H15BFNO2 Formula Weight: 247.07 Physical Form: Solid

Formula: C₁₃H₁₅BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11506358
CAS Number: 1218791-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR