Boc-Arg(NO₂)-OH 98%

Supplier: Thermo Fisher Scientific

H62339.14 H62339.22
H62339.14EA 102 EUR
H62339.14 H62339.22
Boc-Arg(NO₂)-OH 98%
Boc-Arg(NO₂)-OH

Purity: 98% CAS No.: 2188-18-3 Molecular Formula: C11H21N5O6 Formula Weight: 319.31 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 2015163 MDL No.: MFCD00065556

Formula: C₁₁H₂₁N₅O₆
MDL Number: MFCD00065556
CAS Number: 2188-18-3
EINECS: 218-580-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR