3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

H62676.MDEA 0 EUR
H62676.MD
3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester 96%
3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester

Purity: 96% CAS No.: 1192548-05-2 Molecular Formula: C13H15BF4O2 Formula Weight: 290.06 Physical Form: Solid MDL No.: MFCD16996330

Formula: C₁₃H₁₅BF₄O₂
Melting Pt: 34…36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16996330
CAS Number: 1192548-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR