4-Fluoro-3-nitrophenylboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

Synonyms: 2-(4-Fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H62720.03 H62720.06
H62720.03EA 0 EUR
H62720.03 H62720.06
4-Fluoro-3-nitrophenylboronic acid pinacol ester 96%
4-Fluoro-3-nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BFNO₄
MW: 267.06 g/mol
Melting Pt: 64…65 °C
MDL Number: MFCD12026073
CAS Number: 939968-60-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR