4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane 96%

Supplier: Thermo Fisher Scientific

H62764.MD H62764.03
H62764.MDEA 0 EUR
H62764.MD H62764.03
4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane 96%
4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane

Purity: 96% CAS No.: 475250-57-8 Molecular Formula: C17H21BO2 Formula Weight: 268.16 Physical Form: Solid Appearance: White to off-white MDL No.: MFCD22419278

Formula: C₁₇H₂₁BO₂
Storage Temperature: Ambient
MDL Number: MFCD22419278
CAS Number: 475250-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR