Fmoc-O-(benzylphospho)-L-threonine 97%

Supplier: Thermo Fisher Scientific

Synonyms: N-Fmoc-O-benzylphospho-L-threonine, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)(hydroxy)phosphoryl)oxy)butanoic acid, Fmoc-Thr(PO(OBzl)OH)-OH, Fmoc-Thr(HPO3Bzl)-OH

H63006.MD H63006.03
H63006.MDEA 95.2 EUR
H63006.MD H63006.03
Fmoc-O-(benzylphospho)-L-threonine 97%
Fmoc-O-(benzylphospho)-L-threonine

Purity: 97% CAS No.: 175291-56-2 Molecular Formula: C26H26NO8P Formula Weight: 511.46 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Moisture Sensitive Beilstein Reference: 7557104 MDL No.: MFCD00797870

Formula: C₂₆H₂₆NO₈P
MDL Number: MFCD00797870
CAS Number: 175291-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR