5-bromoquinolin-6-amine 97%

Supplier: Thermo Fisher Scientific
Warning

H63150.03 H63150.06
H63150.03EA 199 EUR
H63150.03 H63150.06
5-bromoquinolin-6-amine 97%
5-bromoquinolin-6-amine

Purity: 97% CAS No.: 50358-42-4 Molecular Formula: C9H7BrN2 Formula Weight: 223.07 Physical Form: Solid Appearance: Yellow to brown MDL No.: MFCD00234503

Formula: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD00234503
CAS Number: 50358-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR