3,4-Xylylacetic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3,4-Dimethylphenylacetic acid

H63178.03 H63178.06
H63178.03EA 51.5 EUR
H63178.03 H63178.06
3,4-Xylylacetic acid 98%
3,4-Xylylacetic acid

Purity: 98% CAS No.: 17283-16-8 Molecular Formula: C10H12O2 Formula Weight: 164.2 Physical Form: Solid MDL No.: MFCD02664684

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664684
CAS Number: 17283-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR