tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 97%

Supplier: Thermo Fisher Scientific
Warning

H63191.03 H63191.06
H63191.03EA 119 EUR
H63191.03 H63191.06
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 97%
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate

Purity: 97% CAS No.: 170911-92-9 Molecular Formula: C15H23N3O2 Formula Weight: 277.36 Physical Form: Solid MDL No.: MFCD04115065

Formula: C₁₅H₂₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04115065
CAS Number: 170911-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR