2-chloro-5-iodopyrimidine 95%

Supplier: Thermo Fisher Scientific
Warning

H63423.03 H63423.06
H63423.03EA 75.7 EUR
H63423.03 H63423.06
2-chloro-5-iodopyrimidine 95%
2-chloro-5-iodopyrimidine

Purity: 95% CAS No.: 32779-38-7 Molecular Formula: C4H2ClIN2 Formula Weight: 240.43 Storage Temperature: -30°C to -10°C Physical Form: Solid Sensitivity: Light Sensitive Appearance: Off-white MDL No.: MFCD08062390

Formula: C₄H₂ClIN₂
MDL Number: MFCD06200210
CAS Number: 32779-38-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR