N-Benzoyl-L-tyrosine ethyl ester 98%

Supplier: Thermo Fisher Scientific

H63434.06 H63434.14
H63434.06EA 55.1 EUR
H63434.06 H63434.14
N-Benzoyl-L-tyrosine ethyl ester 98%
N-Benzoyl-L-tyrosine ethyl ester

Purity: 98% CAS No.: 3483-82-7 Molecular Formula: C18H19NO4 Formula Weight: 313.35 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 118-121° Beilstein Reference: 2223819 MDL No.: MFCD00002388

Formula: C₁₈H₁₉NO₄
Melting Pt: 118…121 °C
Storage Temperature: Freezer
MDL Number: MFCD00002388
CAS Number: 3483-82-7
EINECS: 222-469-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR