Fmoc-4-chloro-L-phenylalanine 98%

Supplier: Thermo Fisher Scientific

Synonyms: Fmoc-4-chloro-Phe-OH, N-Fmoc-4-chloro-L-phenylalanine, Fmoc-Phe(4-Cl)-OH

H63448.03 H63448.06
H63448.03EA 108 EUR
H63448.03 H63448.06
Fmoc-4-chloro-L-phenylalanine 98%
Fmoc-4-chloro-L-phenylalanine

Purity: 98% CAS No.: 175453-08-4 Molecular Formula: C24H20ClNO4 Formula Weight: 421.87 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 9437838 MDL No.: MFCD00080281

Formula: C₂₄H₂₀ClNO₄
MDL Number: MFCD00080281
CAS Number: 175453-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR