O-tert-Butyl-L-serine 98%

Supplier: Thermo Fisher Scientific

H63812.03 H63812.06
H63812.03EA 60.9 EUR
H63812.03 H63812.06
O-tert-Butyl-L-serine 98%
O-tert-Butyl-L-serine

Purity: 98% CAS No.: 18822-58-7 Molecular Formula: C7H15NO3 Formula Weight: 161.2 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 220° dec Beilstein Reference: 2500118 Optical Rotation: -16°(c=1 in water) MDL No.: MFCD00066088

Formula: C₇H₁₅NO₃
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00066088
CAS Number: 18822-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR