N-Methyl-3-(trifluoromethyl)benzylamine 95%

Supplier: Thermo Fisher Scientific
Danger

H63888.03 H63888.06
H63888.03EA 105 EUR
H63888.03 H63888.06
N-Methyl-3-(trifluoromethyl)benzylamine 95%
N-Methyl-3-(trifluoromethyl)benzylamine

Purity: 95% CAS No.: 90390-07-1 Molecular Formula: C9H10F3N Formula Weight: 189.18 Physical Form: Oil Appearance: Colourless UN No.: UN2735 MDL No.: MFCD04115410

Formula: C₉H₁₀F₃N
Storage Temperature: Ambient
MDL Number: MFCD04115410
CAS Number: 90390-07-1
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR