N,N'-Dicarbobenzoxy-L-cystine 98%

Supplier: Thermo Fisher Scientific

H63917.03 H63917.06 H63917.14
H63917.03EA 0 EUR
H63917.03 H63917.06 H63917.14
N,N'-Dicarbobenzoxy-L-cystine 98%
N,N'-Dicarbobenzoxy-L-cystine

Purity: 98% CAS No.: 6968-11-2 Molecular Formula: C22H24N2O8S2 Formula Weight: 508.56 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 119° Beilstein Reference: 2550640 MDL No.: MFCD00022030

Formula: C₂₂H₂₄N₂O₈S₂
Melting Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022030
CAS Number: 6968-11-2
EINECS: 230-181-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR