5-Bromo-2-chloroaniline 98%

Supplier: Thermo Fisher Scientific
Danger

H64104.06 H64104.14 H64104.22
H64104.06EA 52.1 EUR
H64104.06 H64104.14 H64104.22
5-Bromo-2-chloroaniline 98%
5-Bromo-2-chloroaniline

Purity: 98% CAS No.: 60811-17-8 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Physical Form: Solid Melting Point: 145-150° Beilstein Reference: 2716274 UN No.: UN2811 MDL No.: MFCD06796283

Formula: C₆H₅BrClN
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD06796283
CAS Number: 60811-17-8
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR