N-(Trimethylsilylmethyl)benzylamine 95%

Supplier: Thermo Fisher Scientific
Warning

H64450.14
H64450.14EA 0 EUR
H64450.14
N-(Trimethylsilylmethyl)benzylamine 95%
N-(Trimethylsilylmethyl)benzylamine

Purity: 95% CAS No.: 53215-95-5 Molecular Formula: C11H19NSi Formula Weight: 193.36 Flash Point: 96°(205°F) Physical Form: Liquid Density: 0.88 Refractive Index: 1.4960 Appearance: Pale yellow Boiling Point: 89-90°/5mm Beilstein Reference: 2802488 MDL No.: MFCD00012192

Formula: C₁₁H₁₉NSi
MW: 193.36 g/mol
Boiling Pt: 89…90 °C (5 mmHg)
Density: 0.88
Flash Pt: 96 °C (205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012192
CAS Number: 53215-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR