5-Amino-2-fluoro-4-nitrobenzotrifluoride 98%

Supplier: Thermo Fisher Scientific
Warning

H64928.03 H64928.06
H64928.03EA 119 EUR
H64928.03 H64928.06
5-Amino-2-fluoro-4-nitrobenzotrifluoride 98%
5-Amino-2-fluoro-4-nitrobenzotrifluoride

Purity: 98% CAS No.: 179062-05-6 Molecular Formula: C7H4F4N2O2 Formula Weight: 224.11 Physical Form: Solid Appearance: Off-white Beilstein Reference: 14360199 UN No.: UN2811 MDL No.: MFCD03094267

Formula: C₇H₄F₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03094267
CAS Number: 179062-05-6
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR