2-Amino-5-chlorophenol 97+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Chloro-2-hydroxyaniline

H64978.14 H64978.22
H64978.14EA 95.7 EUR
H64978.14 H64978.22
2-Amino-5-chlorophenol 97+%
2-Amino-5-chlorophenol

Purity: 97+% CAS No.: 28443-50-7 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Physical Form: Powder Appearance: Brown Melting Point: 145-153° Beilstein Reference: 386421 UN No.: UN2811 MDL No.: MFCD02093863

Formula: C₆H₆ClNO
Melting Pt: 145…153 °C
Storage Temperature: Ambient
MDL Number: MFCD02093863
CAS Number: 28443-50-7
EINECS: 249-020-3
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR