tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95%

Supplier: Thermo Fisher Scientific

H66193.03 H66193.06 H66193.14
H66193.03EA 0 EUR
H66193.03 H66193.06 H66193.14
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate

Purity: 95% CAS No.: 286961-14-6 Molecular Formula: C16H30BNO5 Formula Weight: 327.23 Physical Form: Solid MDL No.: MFCD03840345

Formula: C₁₆H₂₈BNO₄
Storage Temperature: Ambient
MDL Number: MFCD03840345
CAS Number: 286961-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR