Tetramethyl orthocarbonate 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tetramethylorthocarbonate

H66617.03 H66617.06
H66617.03EA 0 EUR
H66617.03 H66617.06
Tetramethyl orthocarbonate 95%
Tetramethyl orthocarbonate

Purity: 95% CAS No.: 1850-14-2 Molecular Formula: C5H12O4 Formula Weight: 136.15 Flash Point: 22°(72°F) Physical Form: Liquid Density: 1.017 Refractive Index: 1.3850 Sensitivity: Moisture Sensitive Appearance: Colourless Melting Point: -5° Boiling Point: 114° Beilstein Reference: 1737682 UN No.: UN3272 MDL No.: MFCD00008473

Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 114 °C
Melting Pt: –5 °C
Density: 1.017
Flash Pt: 22 °C (72 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008473
CAS Number: 1850-14-2
EINECS: 217-438-5
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR