Caronic anhydride 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3,3-Dimethyl-1,2-cyclopropanedicarboxylic Anhydride, 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione

H66892.14 H66892.22
H66892.14EA 0 EUR
H66892.14 H66892.22
Caronic anhydride 98%
Caronic anhydride

Purity: 98% CAS No.: 67911-21-1 Molecular Formula: C7H8O3 Formula Weight: 140.14 Physical Form: Solid Sensitivity: Moisture Sensitive Appearance: White Melting Point: 53-57° Boiling Point: 137°/14mm Beilstein Reference: 117391 MDL No.: MFCD09751198

Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 137 °C (14 mmHg)
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD09751198
CAS Number: 67911-21-1
EINECS: 614-164-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR