Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Supplier: Hach
1206499 1417099 2076032 2105569 2122332 2168049 2212129 2212242 2297255 230149 179032 179049 2141299 2105769 2319900 2244700 2244500 2458200 2459200 2293666 2244100 2070042 2070142 2070242 2446600 2241732 2241832 2229726 2257169 2658300 2257700 2243400 2243900 2459300 2459100 2430200 2603100 2603101 2277800 2352449 2352512 2375000 2368531 2277924 2278024 2429300 2401066 2401268 2544800 2429700 2244600 2604299 2319800 2563000 2459000 2242300 2603300 2603700 2296600 2449400 199526 2651600 2658448 2293566 2770900 2790800 2168042 44449 2076053 2678500 2802246 HPT240 2122326 2678600 2076049 2754048 2947553 HPT310 2368749 2368964 2368655 2368855 2105728 2527025 2658825
HACH1206499EA 138 EUR
HACH1206499 705-0558 705-0083 705-0085 HACH2122332 HACH2168049 HACH2212129 HACH2212242 HACH2297255 705-2035 705-0077 HACH179049 705-0100 705-0088 705-0121 705-2032 705-0110 705-0139 705-0141 HACH2293666 705-0108 HACH2070042 HACH2070142 HACH2070242 705-0137 HACH2241732 HACH2241832 HACH2229726 HACH2257169 HACH2658300 HACH2257700 HACH2243400 HACH2243900 705-0142 705-0140 705-0134 HACH2603100 HACH2603101 705-0116 HACH2352449 HACH2352512 705-0123 HACH2368531 458-0045 458-0046 705-0130 HACH2401066 HACH2401268 705-0152 705-2036 705-2031 HACH2604299 705-0120 HACH2563000 HACH2459000 HACH2242300 705-0156 705-0157 705-2034 705-0560 HACH199526 705-0169 HACH2658448 HACH2293566 705-2038 HACH2790800 HACH2168042 705-0199 HACH2076053 HACH2678500 HACH2802246 HACHHPT240 HACH2122326 HACH2678600 HACH2076049 HACH2754048 HACH2947553 HACHHPT310 HACH2368749 HACH2368964 HACH2368655 HACH2368855 HACH2105728 HACH2527025 HACH2658825
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Environmental Test Kits
Reagents for photometric analysis either in field conditions or in the laboratory.
PP - Powder pillows - Simple and accurate addition of the reagents from a hermetically sealed aluminium foil pillow - ideal for measurement in difficult conditions.
AV - ACCUVAC - Sealed glass cuvettes containing a portioned amount of reagent. Simply immerse the tip of the accuvac in the sample, break it and the vacuum draws the sample into the cuvette.
IM - Immunoassay method of test.
RL- Rapid liquid test kit - Often used for trace level detection (e.g ultra low silica or total chlorine) and in conjunction with a flow cell when there is a high sample throughput.
RS - Reagent solution - Standard chemical reagents supplied in a bottle.
Caution: * Suitable only for the DR 5000 UV-Vis Instrument.