Sodium standard solutions (NIST)
Supplier: Hach
2318153
HACH2318153EA 76.9 EUR
HACH2318153
Sodium standard solutions (NIST)
Standards Cationic Standards Sodium Standards
For accuracy checks and instrument calibrations in sodium determinations.
Caution: 100 mg/l = 100 µg/ml