pH buffer, single-use, SINGLET™
Supplier: Hach
2770020 2770120 2770220 2769820 2769920
HACH2770020EA 53.9 EUR
HACH2770020 HACH2770120 HACH2770220 HACH2769820 HACH2769920
pH buffer, single-use, SINGLET™
Buffers pH Calibration Buffers
These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.
  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0,02 at 25 °C
  • Traceable to NIST
Order Now
SPECIFICATIONS
Environmentally Preferable