Wastewater inorganics quality control standard
Supplier: Hach
2833249 2833149
HACH2833249EA 181 EUR
HACH2833249 HACH2833149
Wastewater inorganics quality control standard
Standards Environmental Analysis Standards
These mixed standards are ready to use, no dilutions are required. They contain ammonia nitrogen, nitrate nitrogen, COD, phosphate, sulphate and TOC.