Electrode storage bottles
Supplier: Hach
9260300
HACH9260300EA 6 EUR
HACH9260300
Electrode storage bottles
Electrode Holders
Electrode storage bottle is used to store pH electrodes.
  • Rubber grommet in lid ensures tight seal with 12 mm electrodes