Sodium periodate

Supplier: Hach
Danger

98499
HACH98499EA 73.2 EUR
HACH98499
Sodium periodate
Sodium periodate
Formula: NaIO₄
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 175 °C
Density: 3,87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 13,08714

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR