ORP Electrode, sensION+ 5055
Supplier: Hach
LZW5055970002
HACHLZW5055970002EA 332 EUR
HACHLZW5055970002
ORP Electrode, sensION+ 5055
Electrodes Redox Electrodes
sensION+ 5055 is a combination platinum ORP electrode with a polycarbonate body and non refillable gel-electrolyte reference. It is dedicated for use with Hach sensION+ portable pH (mV) meters.
  • Fixed 1 meter cable and MP5 connector
  • Resilient PC body, heavy-duty handle
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimised for field calibration and storage
The 5055 is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.