Electrode cleaning solution, RENOVO.N
Supplier: Hach
S16M001
HACHS16M001EA 27.7 EUR
HACHS16M001
Electrode cleaning solution, RENOVO.N
Electrolytes Electrode Storage Solutions
Mild cleaning solution for electrodes. Ideal for use after measurements in aqueous solutions.