Immersion thermostats
Supplier: THERMO ELECTRON LED
152-0001 1520018 152-0011 153-0011 152-0011 1520011 1530011 1520001B
HAKA152-0001EA 0 EUR
HAKA152-0001 HAKA1520018 HAAK152-0011 HAKA153-0011 HAKA152-0011 HAKA1520011 HAKA1530011 HAKA1520001B
Immersion thermostats
Circulating Thermostats Open Circulator Systems Thermostat Controllers
Designed for ease-of-use and to be energy efficient with powerful pumping and heating capacities for closed loop applications.
  • Self-tuning PID controller for optimised temperature control
  • Five programmable set point temperatures
  • RTA (Real Temperature Adjustment) for calibration
  • Two levels of pump speed adjustment to increase flow or bath agitation
  • Three languages (English, German or French)
  • Change digital display resolution between 0,1 and 0,01 and between °C — °F — °K
  • Acoustic and optical alarm
  • Auto-restart feature after power failure
These basicMore Product Information